MilliporeSigma
  • Antihormonal agents as a strategy to improve the effect of chemo-radiation in cervical cancer: in vitro and in vivo study.
Antihormonal agents as a strategy to improve the effect of chemo-radiation in cervical cancer: in vitro and in vivo study.
BMC cancer (2015-01-28)
Mariana Segovia-Mendoza, Rafael Jurado, Roser Mir, Luis A Medina, Heriberto Prado-Garcia, Patricia Garcia-Lopez
ABSTRACT
Over the past few years, the concurrent use of cisplatin-based chemotherapy and radiation therapy has dramatically improved the local response and increased overall survival in early-stage cervical cancer. However, for the advanced stages of the disease this standard treatment has proved insufficient. We investigated the capacity of Mifepristone and ICI 182,780, which are anti-progestin and anti-estrogen drugs, respectively, to act as chemo-radiosensitizing agents in cervical cancer cells and cervix xenografts. The effect of chemo-radiation alone or combined with Mifepristone or ICI 182,780 was evaluated in HeLa cells and with tumor growth in cervix xenografts. After concomitant chemo-radiotherapy, the effect of each of these antihormonal agents on apoptosis (determined by Annexing V assay) and the cell cycle phases were determined by flow cytometry. The expression of angiogenic factor VEGF in tumor samples was determined using quantitative RT-PCR analysis of VEGF gene expression. Compared to radiation alone or radiation/cisplatin therapy, there was significantly higher cytotoxicity and a greater antitumoral effect with the combined application of radiation/cisplatin and Mifepristone or ICI 182,780. Analyses of the apoptosis and cell cycle demonstrated changes only with ICI, not with Mifepristone, when was applied in combination with radiation/cisplatin. The analysis of VEGF mRNA expression levels in tumors at the end of the study demonstrated a significant inhibition, compared to radiation only or the radiation/cisplatin treatment, after concurrent chemo-radiotherapy and each one of the antihormonal drugs. Mifepristone and ICI 182,780 may be potentially promising chemo-radiosensitizing compounds to be used in combination with ionizing irradiation and cisplatin in the treatment of patients with advanced cervical cancer.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Chloroform, analytical standard
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis