  • Search Results
  • Monitoring urinary metabolites resulting from sulfur mustard exposure in rabbits, using highly sensitive isotope-dilution gas chromatography-mass spectrometry.
Monitoring urinary metabolites resulting from sulfur mustard exposure in rabbits, using highly sensitive isotope-dilution gas chromatography-mass spectrometry.
Analytical and bioanalytical chemistry (2014-06-14)
Zhiyong Nie, Yajiao Zhang, Jia Chen, Ying Lin, Bidong Wu, Yuan Dong, Jianlin Feng, Qin Liu, Jianwei Xie
ABSTRACT
A highly sensitive method for the determination of sulfur mustard (SM) metabolites thiodiglycol (TDG) and thiodiglycol sulfoxide (TDGO) in urine was established and validated using isotope-dilution negative-ion chemical ionization (NICI) gas chromatography-mass spectrometry (GC-MS). TDGO in the samples was reduced with TiCl3, and then determined together with TDG as a single analyte. The sample preparation procedures, including two solid-phase-extraction (SPE) clean-up steps, were optimized to improve the sensitivity of the method. The limits of detection (LOD) for both TDG and TDG plus TDGO (TDG + TDGO) were 0.1 ng mL(-1), and the limits of quantitation (LOQ) for both were 0.3 ng mL(-1). The method was used in a rabbit cutaneous SM exposure model. Domestic rabbits were exposed to neat liquid SM at three dosage levels (0.02, 0.05, and 0.15 LD50), and the urinary excretion of four species of hydrolysis metabolites, namely free TDG, free plus conjugated TDG (total TDG), free TDG + TDGO, and free plus conjugated TDG + TDGO (total TDG + TDGO), was evaluated to investigate the metabolic processes. The total urinary excretion profiles of the metabolites, including the peak time, time window, and dose-response and time-response relationships, were clarified. The results revealed that the concentrations of TDG and TDG + TDGO in the urine increased quickly and then decreased rapidly in the first two days after SM exposure. The cumulative amount of total TDG + TDGO excreted in urine during the first five days accounted for 0.5-1% of the applied dose of SM. It is also concluded that TDG and TDGO in urine existed mainly in free form, the levels of glucuronide and of sulfate conjugates of TDG or TDGO were very low, and most hydrolysis metabolites were present in the oxidized form (TDGO). The study indicates that the abnormal increase of TDG and TDGO excretion levels can be used as a diagnostic indicator and establishes a reference time-window for retrospective analysis and sampling after SM exposure.
Sigma-Aldrich
Pyridine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
2,2,3,3,4,4,4-Heptafluorobutyl acrylate, 97%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
2,3,4,5,6-Pentafluorobenzoyl chloride, 99%
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Pyridine, ≥99%
Supelco
Pyridine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Heptafluorobutyric anhydride, for GC derivatization, LiChropur, ≥99.0%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Heptafluorobutyric anhydride, ≥98%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Supelco
Heptafluorobutyric anhydride, for GC derivatization, LiChropur, ≥99.0%
Supelco
Triethylamine, analytical standard