MilliporeSigma
  • Photodiode array to charged aerosol detector response ratio enables comprehensive quantitative monitoring of basic drugs in blood by ultra-high performance liquid chromatography.
Photodiode array to charged aerosol detector response ratio enables comprehensive quantitative monitoring of basic drugs in blood by ultra-high performance liquid chromatography.
Analytica chimica acta (2015-03-04)
Jenni Viinamäki, Ilkka Ojanperä
ABSTRACT
Quantitative screening for a broad range of drugs in blood is regularly required to assess drug abuse and poisoning within analytical toxicology. Mass spectrometry-based procedures suffer from the large amount of work required to maintain quantitative calibration in extensive multi-compound methods. In this study, a quantitative drug screening method for blood samples was developed based on ultra-high performance liquid chromatography with two consecutive detectors: a photodiode array detector and a corona charged aerosol detector (UHPLC-DAD-CAD). The 2.1 mm × 150 mm UHPLC column contained a high-strength silica C18 bonded phase material with a particle size of 1.8 μm, and the mobile phase consisted of methanol/0.1% trifluoroacetic acid in gradient mode. Identification was based on retention time, UV spectrum and the response ratio from the two detectors. Using historic calibration over a one-month period, the median precision (RSD) of retention times was 0.04% and the median accuracy (bias) of quantification 6.75%. The median precision of the detector response ratio over two orders of magnitude was 12%. The applicable linear ranges were generally 0.05-5 mg L(-1). The method was validated for 161 compounds, including antipsychotics, antidepressants, antihistamines, opioid analgesics, and adrenergic beta blocking drugs, among others. The main novelty of the method was the proven utility of the response ratio of DAD to CAD, which provided the additional identification efficiency required. Unlike with mass spectrometry, the high stability of identification and quantification allowed the use of facile historic calibration.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Supelco
Ethyl acetate, analytical standard
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate
SAFC
Tromethamine
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
USP
Tromethamine, United States Pharmacopeia (USP) Reference Standard
Trometamol, European Pharmacopoeia (EP) Reference Standard
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)