  • Search Results
  • Excess perigestational folic acid exposure induces metabolic dysfunction in post-natal life.
Excess perigestational folic acid exposure induces metabolic dysfunction in post-natal life.
The Journal of endocrinology (2015-02-11)
Elisa Keating, Ana Correia-Branco, João R Araújo, Manuela Meireles, Rita Fernandes, Luísa Guardão, João T Guimarães, Fátima Martel, Conceição Calhau
ABSTRACT
The aim of this study was to understand whether high folic acid (HFA) exposure during the perigestational period induces metabolic dysfunction in the offspring, later in life. To do this, female Sprague-Dawley rats (G0) were administered a dose of folic acid (FA) recommended for pregnancy (control, C, 2 mg FA/kg of diet, n=5) or a high dose of FA (HFA, 40 mg FA/kg of diet, n=5). Supplementation began at mating and lasted throughout pregnancy and lactation. Body weight and food and fluid intake were monitored in G0 and their offspring (G1) till G1 were 13 months of age. Metabolic blood profiles were assessed in G1 at 3 and 13 months of age (3M and 13M respectively). Both G0 and G1 HFA females had increased body weight gain when compared with controls, particularly 22 (G0) and 10 (G1) weeks after FA supplementation had been stopped. G1 female offspring of HFA mothers had increased glycemia at 3M, and both female and male G1 offspring of HFA mothers had decreased glucose tolerance at 13M, when compared with matched controls. At 13M, G1 female offspring of HFA mothers had increased insulin and decreased adiponectin levels, and G1 male offspring of HFA mothers had increased levels of leptin, when compared with matched controls. In addition, feeding of fructose to adult offspring revealed that perigestational exposure to HFA renders female progeny more susceptible to developing metabolic unbalance upon such a challenge. The results of this work indicate that perigestational HFA exposure the affects long-term metabolic phenotype of the offspring, predisposing them to an insulin-resistant state.
Product Description
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Supelco
D-(−)-Fructose, analytical standard, analytical standard for fructose assay kit, for use with enzymatic assay kit FA20
Sigma-Aldrich
D-(−)-Fructose, BioXtra
Sigma-Aldrich
D-(−)-Fructose, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(−)-Fructose, ≥99%
Sigma-Aldrich
D-(−)-Fructose, meets USP testing specifications
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Millipore
D-(−)-Fructose, ≥99.0% (HPLC), suitable for microbiology
Sigma-Aldrich
D-(−)-Fructose, tested according to Ph. Eur.
Supelco
Fructose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Fructose, European Pharmacopoeia (EP) Reference Standard
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
USP
Fructose, United States Pharmacopeia (USP) Reference Standard
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Folic acid, Pharmaceutical Secondary Standard; Certified Reference Material