  • Search Results
  • Hybrid organic-inorganic colloidal composite 'sponges' via internal crosslinking.
Hybrid organic-inorganic colloidal composite 'sponges' via internal crosslinking.
Small (Weinheim an der Bergstrasse, Germany) (2014-11-11)
Bradley Duncan, Ryan F Landis, Huda A Jerri, Valery Normand, Daniel Benczédi, Lahoussine Ouali, Vincent M Rotello
ABSTRACT
An effective method for the generation of hybrid organic-inorganic nanocomposite microparticles featuring controlled size and high structural stability is presented. In this process, an oil-in-water Pickering emulsion is formed using hydrophilic amine-functionalized silica nanoparticles. Covalent modification using a hydrophobic maleic anhydride copolymer then alters nanoparticle wettability during crosslinking, causing a core-shell to nanocomposite structural reorganization of the assemblies. The resulting porous nanocomposites maintain discrete microparticle structures and retain payloads in their oil phase even when incubated in competitive solvents such as ethanol.
Sigma-Aldrich
Ammonium hydroxide solution, puriss., 30-33% NH3 in H2O
Sigma-Aldrich
Ammonium hydroxide solution, puriss., meets analytical specification of Ph. Eur., 25-30% NH3 basis
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a. plus, ≥25% NH3 in H2O
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a., reag. ISO, reag. Ph. Eur., ~25% NH3 basis
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Diphenyl ether, ReagentPlus®, ≥99%
Sigma-Aldrich
Diphenyl ether, ReagentPlus®, 99%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Diphenyl ether, ≥99%, FG
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Supelco
Diphenyl ether, Selectophore, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)