MilliporeSigma
5-membered cyclic hydroxamic acids as HDAC inhibitors.
Journal of enzyme inhibition and medicinal chemistry (2014-06-19)
Ilze Mutule, Diana Borovika, Elina Rozenberga, Nadezhda Romanchikova, Raivis Zalubovskis, Irina Shestakova, Peteris Trapencieris
ABSTRACT
The new histone deacylases inhibitors (HDACi) were synthesized in the class of 5-membered cyclic hydroxamic acids (5-CHA), showing medium size CHA as a new Zn-binding group. New reaction sequence was proposed for the synthesis of 5-membered alkylidene-cyclic-hydroxamic acids starting from butyrolactone. Compound 10c showed low µM activity on HeLa cell extracts. From these results, cyclic hydroxamic acids will be further investigated to find more potent compounds.
Sigma-Aldrich
Mercury, electronic grade, 99.9999% trace metals basis
Sigma-Aldrich
Mercury, ≥99.99% trace metals basis
Sigma-Aldrich
Mercury, ACS reagent, 99.9995% trace metals basis