MilliporeSigma
  • Epithelial cell adhesion molecule-positive human hepatic neoplastic cells: development of combined hepatocellular-cholangiocarcinoma in mice.
Epithelial cell adhesion molecule-positive human hepatic neoplastic cells: development of combined hepatocellular-cholangiocarcinoma in mice.
Journal of gastroenterology and hepatology (2014-08-05)
Sachiko Ogasawara, Jun Akiba, Masamichi Nakayama, Osamu Nakashima, Takuji Torimura, Hirohisa Yano
ABSTRACT
Human combined hepatocellular-cholangiocarcinoma (CHC) expresses several hepatic stem/progenitor cell (HSPC) markers, suggesting this neoplasm originates from HSPCs. We examined the significance of HSPC marker in CHC using a human CHC cell line. We used a human CHC cell line (KMCH-1) previously established in our laboratory. The original tumor was classified as CHC, showing areas of typical hepatocellular carcinoma (HCC) and cholangiocarcinoma (ChC). We examined the expression of HSPC markers and hepatocyte markers in KMCH-1 by flow cytometry (FCM) and quantitative real-time polymerase chain reaction. EpCAM(+) and EpCAM(-) KMCH-1 cells were isolated. Subsequently, their morphological features, HSPC marker expression, and biological characteristics were examined in vitro and in vivo. FCM showed expression of EpCAM, K7, K19, and ABCG2 in KMCH-1, with various degrees. EpCAM(+) cells expressed K19 mRNA, but did not express α-fetoprotein (AFP). In contrast, EpCAM(-) cells expressed AFP mRNA, but did not express K19. EpCAM(+) cells produced both EpCAM(+) and EpCAM(-) cells, but EpCAM(-) cells produced only EpCAM(-) cells in vitro. EpCAM(+) cells showed higher tumorigenicity and formed larger tumors than EpCAM(-) cells. Inoculation of EpCAM(+) and EpCAM(-) cells produced both ChC and HCC-like component and HCC-like component only, respectively. It is speculated that some CHCs may originate from EpCAM(+) neoplastic cells, and that these cells may affect malignant behavior and progression in such CHCs.
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard