MilliporeSigma
  • Morphology and gene expression profile of the submandibular gland of androgen-receptor-deficient mice.
Morphology and gene expression profile of the submandibular gland of androgen-receptor-deficient mice.
Archives of oral biology (2014-12-02)
Kannika Adthapanyawanich, Tewarat Kumchantuek, Hiroki Nakata, Miyuki Yamamoto, Tomohiko Wakayama, Takumi Nishiuchi, Shoichi Iseki
ABSTRACT
In the submandibular gland (SMG) of mice, the granular convoluted tubule (GCT) develops preferentially in males dependent on androgens. To clarify the molecular mechanism of androgen action in SMG, we examined the SMG of mice deficient for the androgen receptor (ARKO). The morphological features and gene expression in the SMG of control and ARKO mice with or without hormone treatments were analysed by immunohistochemistry, DNA microarray, and RT-PCR. The development of GCT and expression of GCT-specific products such as NGF were even lower in ARKO male SMG than in control female SMG. The administration of androgens to ARKO males had no effect on SMG, whereas the administration of thyroid hormone (T4) caused the extensive conversion of striated duct cells to GCT cells with the increase of NGF mRNA. Gene expression profiles in control and ARKO male SMG were analysed by DNA microarrays, and genes with higher or lower expression in ARKO male SMG were determined. They were then classified into groups according to their responsiveness to the administration of dihydotestosterone (DHT) or T4 to ARKO males. RT-PCR revealed that, while no gene was responsive to DHT, expression of many genes was up- or down-regulated by T4. These results confirmed that GCT cell differentiation induced by androgens is dependent on the classical androgen receptor (AR), whereas that by T4 is independent of AR. Differential reactivity of genes to androgens and thyroid hormone in ARKO mice may shed light on the mechanism of androgen action in the SMG.
Product Description
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Testosterone, ≥98%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Testosterone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard