MilliporeSigma
Controlling the misuse of cobalt in horses.
Drug testing and analysis (2014-09-27)
Emmie N M Ho, George H M Chan, Terence S M Wan, Peter Curl, Christopher M Riggs, Michael J Hurley, David Sykes
ABSTRACT
Cobalt is a well-established inducer of hypoxia-like responses, which can cause gene modulation at the hypoxia inducible factor pathway to induce erythropoietin transcription. Cobalt salts are orally active, inexpensive, and easily accessible. It is an attractive blood doping agent for enhancing aerobic performance. Indeed, recent intelligence and investigations have confirmed cobalt was being abused in equine sports. In this paper, population surveys of total cobalt in raceday samples were conducted using inductively coupled plasma mass spectrometry (ICP-MS). Urinary threshold of 75 ng/mL and plasma threshold of 2 ng/mL could be proposed for the control of cobalt misuse in raceday or in-competition samples. Results from administration trials with cobalt-containing supplements showed that common supplements could elevate urinary and plasma cobalt levels above the proposed thresholds within 24 h of administration. It would therefore be necessary to ban the use of cobalt-containing supplements on raceday as well as on the day before racing in order to implement and enforce the proposed thresholds. Since the abuse with huge quantities of cobalt salts can be done during training while the use of legitimate cobalt-containing supplements are also allowed, different urinary and plasma cobalt thresholds would be required to control cobalt abuse in non-raceday or out-of-competition samples. This could be achieved by setting the thresholds above the maximum urinary and plasma cobalt concentrations observed or anticipated from the normal use of legitimate cobalt-containing supplements. Urinary threshold of 2000 ng/mL and plasma threshold of 10 ng/mL were thus proposed for the control of cobalt abuse in non-raceday or out-of-competition samples.
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Isoamyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Isoamyl alcohol, ≥98%, FG
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
3-Methylbutanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Supelco
3-Methylbutanol, analytical standard
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
3-Methylbutanol, BioReagent, for molecular biology, ≥98.5%
Sigma-Aldrich
Indole-3-acetic acid sodium salt, BioReagent, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Nitric acid, red, fuming, HNO3 >90 %
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Supelco
Citric acid, certified reference material, TraceCERT®
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Supelco
3-Indoleacetic acid, PESTANAL®, analytical standard