MilliporeSigma
Cell growth on pore-graded biomimetic TiO2 bone scaffolds.
Journal of biomaterials applications (2014-11-15)
Benjamin Müller, Janne Elin Reseland, Håvard Jostein Haugen, Hanna Tiainen
ABSTRACT
In order to prevent soft tissue down-growth into osseous defect areas, membranes are used when placing bone graft materials. These membranes still show shortcomings in their performance and applications. In the current study, we choose an approach to integrate micro-porous surface structures into a macro-porous scaffold. Low porous surfaces were fabricated by dip-coatings. Four different material compositions (titanium dioxide, polycaprolactone, polycaprolactone/water, polycaprolactone/β-tricalcium phosphate) were characterised in terms of their appearance, architecture, topographical features and cell response. Titanium dioxide surfaces exhibited rougher and more complex textures, resulting in the highest number of osteosarcoma cells and distinct morphologies in terms of cell spreading. Polycaprolactone-based surfaces showed a smoother topography and enhanced microporosity, but the effect on secretion of the bone markers sclerostin and interleukin-6 from human osteoblasts was lower compared to secretion from cells cultured on titanium dioxide. β-Tricalcium phosphate modification of polycaprolactone did not show any significant improvement regarding cell-material interaction. Nevertheless, surfaces show potential in the mechanical blockage of epithelial and soft tissue cells and may still permit sufficient nutrient transport.
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
β-tri-Calcium phosphate, puriss. p.a., ≥98% β-phase basis (sintered Powder)
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
β-tri-Calcium phosphate, puriss. p.a., ≥98% β-phase basis (unsintered powder)
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
α-Tri-Calcium phosphate, Reagent for transient & stable DNA transfections
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Supelco
Dichloromethane solution, certified reference material, 200 μg/mL in methanol
Supelco
Dichloromethane, analytical standard, stabilized
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine
Supelco
Dichloromethane, ≥99.9%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
Dichloromethane, Selectophore, ≥99.5%
Supelco
L-Glutamine, certified reference material, TraceCERT®
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Methylene chloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source