  • Search Results
  • Three-layer poly(methyl methacrylate) microsystem for analysis of lysosomal enzymes for diagnostic purposes.
Three-layer poly(methyl methacrylate) microsystem for analysis of lysosomal enzymes for diagnostic purposes.
Analytica chimica acta (2014-12-04)
Radoslaw Kwapiszewski, Karina Kwapiszewska, Jörg P Kutter, Zbigniew Brzozka
ABSTRACT
Lysosomal storage diseases are chronic, progressive and typically have a devastating impact on the patient and the family. The diagnosis of these diseases is still a challenge, however, even for trained specialists. Accurate diagnostic methods and high-throughput tools that could be readily incorporated into existing screening laboratories are urgently required. We propose a new method for measuring the activity of lysosomal enzymes using a microfluidic device. The principle of the method is the fluorometric determination of a protonated form of 4-methylumbelliferone directly in the enzymatic mixture. Compared to the standard diagnostic protocols, the method eliminates the necessity to add alkaline buffer to stop the enzymatic reaction, and thus, the number of analytical steps is reduced. The system allows for on-chip short-term incubation of the enzymatic reagents, leading to a much simplified analytical procedure and a significantly shortened processing time. We measured the activity of β-galactosidase in RPMI-1788 human B lymphocytes and in isolated leukocytes from healthy adults. The method shows a good agreement with the standard diagnostic method. The agreement was confirmed by statistical analysis including construction of a Bland-Altman plot. The approach presented can be an alternative for the currently used diagnostic procedures. The method developed has a potential for the implementation into complex microfluidic devices thus becoming a powerful tool for a high-throughput and multiplex screening of newborns.
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
4-Methylumbelliferone, ≥98%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Glycine, 99%, FCC
SAFC
Glycine
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)