MilliporeSigma
  • Redox-responsive polymer-drug conjugates based on doxorubicin and chitosan oligosaccharide-g-stearic acid for cancer therapy.
Redox-responsive polymer-drug conjugates based on doxorubicin and chitosan oligosaccharide-g-stearic acid for cancer therapy.
Molecular pharmaceutics (2015-03-10)
Yigang Su, Yingwen Hu, Yongzhong Du, Xuan Huang, Jiabei He, Jian You, Hong Yuan, Fuqiang Hu
ABSTRACT
Here, a biodegradable polymer-drug conjugate of doxorubicin (DOX) conjugated with a stearic acid-grafted chitosan oligosaccharide (CSO-SA) was synthesized via disulfide linkers. The obtained polymer-drug conjugate DOX-SS-CSO-SA could self-assemble into nanosized micelles in aqueous medium with a low critical micelle concentration. The size of the micelles was 62.8 nm with a narrow size distribution. In reducing environments, the DOX-SS-CSO-SA could rapidly disassemble result from the cleavage of the disulfide linkers and release the DOX. DOX-SS-CSO-SA had high efficiency for cellular uptake and rapidly released DOX in reductive intracellular environments. In vitro antitumor activity tests showed that the DOX-SS-CSO-SA had higher cytotoxicity against DOX-resistant cells than free DOX, with reversal ability up to 34.8-fold. DOX-SS-CSO-SA altered the drug distribution in vivo, which showed selectively accumulation in tumor and reduced nonspecific accumulation in hearts. In vivo antitumor studies demonstrated that DOX-SS-CSO-SA showed efficient suppression on tumor growth and relieved the DOX-induced cardiac injury. Therefore, DOX-SS-CSO-SA is a potential drug delivery system for safe and effective cancer therapy.
Product Description
Supelco
Pyrene, certified reference material, TraceCERT®
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Supelco
Pyrene, analytical standard
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
Pyrene, puriss. p.a., for fluorescence, ≥99.0% (GC)
Pyrene, BCR®, certified reference material
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Doxorubicin hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
1,2-Dichloroethane, analytical standard
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O