  • Search Results
  • Probing the size limit for nanomedicine penetration into Burkholderia multivorans and Pseudomonas aeruginosa biofilms.
Probing the size limit for nanomedicine penetration into Burkholderia multivorans and Pseudomonas aeruginosa biofilms.
Journal of controlled release : official journal of the Controlled Release Society (2014-08-16)
Katrien Forier, Anne-Sophie Messiaen, Koen Raemdonck, Hans Nelis, Stefaan De Smedt, Jo Demeester, Tom Coenye, Kevin Braeckmans
ABSTRACT
Encapsulation of antibiotics into nanoparticles is a potential strategy to eradicate biofilms. To allow further optimization of nanomedicines for biofilm eradication, the influence of the nanoparticle size on the penetration into dense biofilm clusters needs to be investigated. In the present study, the penetration of nanoparticles with diameters ranging from 40 to 550 nm into two biofilms, Burkholderia multivorans LMG 18825 and Pseudomonas aeruginosa LMG 27622, was evaluated using confocal microscopy. Through image analysis, the percentage of particles able to penetrate into dense biofilm clusters was calculated. The size cut off for optimal penetration into biofilm clusters was located around 100-130 nm for both biofilms. The mesh size of the biofilm matrix and the size of the channels in between the bacteria of the clusters are two factors which likely play a role in the exclusion of the larger particles. For B. multivorans, a sharp drop in the penetration into the clusters is seen for particles larger than 130 nm while for P. aeruginosa, a more gradual decrease in penetration could be observed. The overall penetration of the nanoparticles was slightly lower for P. aeruginosa than for B. multivorans. Based on these results, it could be concluded that nanocarriers of about 100 nm and smaller are good candidates to improve the treatment of chronic pulmonary biofilms in CF patients. Furthermore, the confocal microscopy method demonstrated here is a useful tool to assess the penetration of nanomedicines in biofilm clusters. Such information is important to optimize nanomedicine formulations for the treatment of biofilm infections.
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, commercial grade, powder
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, BioXtra
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, crystalline
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, purum, ≥98.0% (AT)
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, ≥99.0% (AT)
SAFC
Cholesterol, Plant-Derived, SyntheChol®, NF, Ph. Eur., JP
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Supelco
Chloroform, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)