  • Search Results
  • Exploratory analysis of urinary tract infection using a GC-APCI-MS platform.
Exploratory analysis of urinary tract infection using a GC-APCI-MS platform.
The Analyst (2015-03-03)
Tiziana Pacchiarotta, Rico J Derks, Ekaterina Nevedomskaya, Willize van der Starre, Jaap van Dissel, André Deelder, Oleg A Mayboroda
ABSTRACT
Urinary tract infection (UTI) is among the most common bacterial infections worldwide. The understanding of the physiological mechanisms affected by UTI may need modern integrative '-omics' technologies, and metabolomics in particular. Here we present the first GC-APCI-MS-based explorative metabolomics study of UTI, using MS and FID detectors simultaneously. This provides high quality mass spectral data as well as semi-quantitative information demonstrating the feasibility of the GC-APCI-MS platform for non-targeted approaches. The work is part of a bigger project aiming at providing a comprehensive overview of UTI-induced changes in urine. Taking advantage of a fully clinically characterized cohort that offers the possibility of both case-control and longitudinal modelling, we can define UTI-induced change as a list of urinary metabolites which distinguish E. coli UTI patients from the subjects with no signs of an active infection. The list of molecular descriptors includes compounds related to bacterial activity such as lactic acid and lactose while other molecules show an association with the physiological status (inositol, citric acid).
Sigma-Aldrich
Pyridine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Acetic anhydride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99% (GC)
Supelco
3-(3-Hydroxyphenyl)-3-hydroxypropionic acid, analytical standard
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Supelco
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, synthesis grade
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material