  • Search Results
  • Early outcome prediction on 18F-fluorocholine PET/CT in metastatic castration-resistant prostate cancer patients treated with abiraterone.
Early outcome prediction on 18F-fluorocholine PET/CT in metastatic castration-resistant prostate cancer patients treated with abiraterone.
Oncotarget (2014-12-17)
Ugo De Giorgi, Paola Caroli, Salvatore L Burgio, Cecilia Menna, Vincenza Conteduca, Emanuela Bianchi, Francesca Fabbri, Elisa Carretta, Dino Amadori, Giovanni Paganelli, Federica Matteucci
ABSTRACT
We investigated the role of 18F-fluorocholine positron emission tomography/computed tomography (FCH-PET/CT) in the early evaluation of abiraterone and outcome prediction in patients with metastatic castration-resistant prostate cancer (CRPC). Forty-three patients with metastatic CRPC progressing after docetaxel received abiraterone 1,000 mg daily with prednisone 5 mg twice daily. Patients were evaluated monthly for serological PSA response and safety. FCH-PET/CT was done at baseline and after 3 to 6 weeks. Univariate and multivariate Cox regression models addressed potential predictors of progression-free survival (PFS) and overall survival (OS). Declines in PSA level of ≥50% were seen in 21 of 43 (49%) patients. Forty-two patients were evaluable for FCH-PET/CT response. FCH-PET/CT bone flare was observed in 4 of 42 (10%) evaluable patients. In univariate analysis, PSA decline and FCH-PET/CT response predicted PFS, while PSA decline and FCH-PET/CT (progression vs non progression) predicted OS. In multivariate analysis, only FCH-PET/CT (progression vs nonprogression) remained significant for PFS and OS (p = 0.022 and p = 0.027, respectively). Early FCH-PET/CT can predict clinical outcome in CRPC beyond PSA response. These data support further studies on FCH-PET/CT for abiraterone monitoring and outcome prediction in patients with CRPC.
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Tantalum(V) ethoxide, packaged for use in deposition systems