  • Search Results
  • Drp1 stabilizes p53 on the mitochondria to trigger necrosis under oxidative stress conditions in vitro and in vivo.
Drp1 stabilizes p53 on the mitochondria to trigger necrosis under oxidative stress conditions in vitro and in vivo.
The Biochemical journal (2014-04-25)
Xing Guo, Hiromi Sesaki, Xin Qi
ABSTRACT
Oxidative-stress-induced necrosis is considered to be one of the main pathological mediators in various neurological disorders, such as brain ischaemia. However, little is known about the mechanism by which cells modulate necrosis in response to oxidative stress. In the present study, we showed that Drp1 (dynamin-related protein 1), a primary mitochondrial fission protein, stabilizes the well-known stress gene p53 and is required for p53 translocation to the mitochondria under conditions of oxidative stress. We found that Drp1 binding to p53 induced mitochondria-related necrosis. In contrast, inhibition of Drp1 hyperactivation by Drp1 siRNA reduced necrotic cell death in cell cultures exposed to oxidative stress. Most significantly, we demonstrated that inhibition of Drp1 by the Drp1 peptide inhibitor P110, which was developed recently by our group, abolished p53 association with the mitochondria and reduced brain infarction in rats subjected to brain ischaemia/reperfusion injury. Taken together, these findings reveal a novel mechanism of Drp1 hyperactivation in the induction of mitochondrial damage and subsequent cell death. We propose that a Drp1 inhibitor such as P110 is a possible therapeutic agent for diseases in which hyperactivated Drp1 contributes to the pathology.
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Rabbit anti-p53 Antibody, Affinity Purified
Sigma-Aldrich
Anti-TP53 (Ab-33) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-TP53 (Ab-18) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-TP53 (Ab-9) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
3-Nitropropionic acid, ≥97%
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Anti-TP53 (Ab-46) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-TP53 (Ab-37) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Anti-p53 antibody produced in sheep, fractionated antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-p53 antibody, Mouse monoclonal, clone BP53-12, ascites fluid
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
Anti-p53 antibody, Mouse monoclonal, clone PAb1801, purified from hybridoma cell culture
Sigma-Aldrich
Anti-p53 antibody, Mouse monoclonal, clone DO-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-TP53 antibody produced in rabbit, IgG fraction of antiserum
Sigma-Aldrich
Anti-p53 antibody, Mouse monoclonal, clone DO-7, purified from hybridoma cell culture
Sigma-Aldrich
Anti-TP53 (Ab-315) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-TP53 (Ab-6) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-p53 antibody, Mouse monoclonal, clone DO-2, purified from hybridoma cell culture
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Sigma-Aldrich
Anti-TP53 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-TRP53 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-p53 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL2199, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
ANTI-P53 (S315) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
ANTI-P53 (S15) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution