  • Search Results
  • The non-peptide thrombopoietin receptor agonist eltrombopag stimulates megakaryopoiesis in bone marrow cells from patients with relapsed multiple myeloma.
The non-peptide thrombopoietin receptor agonist eltrombopag stimulates megakaryopoiesis in bone marrow cells from patients with relapsed multiple myeloma.
Journal of hematology & oncology (2015-04-19)
Jee-Yeong Jeong, Michelle S Levine, Nirmalee Abayasekara, Nancy Berliner, Jacob Laubach, Gary J Vanasse
ABSTRACT
Thrombocytopenia is a significant problem in patients with relapsed or refractory multiple myeloma, precipitating a need for supportive platelet transfusions and necessitating decreases in delivered doses of chemotherapy. Eltrombopag is a non-peptide, small molecule thrombopoietin (TPO) receptor agonist that promotes megakaryopoiesis similar to endogenous human TPO and may be an effective agent for thrombocytopenia in this patient population. We examined the effects of eltrombopag on megakaryocyte colony-forming capacity in CD34+ cells in patients with multiple myeloma and investigated its impact on proliferation, viability, and apoptosis in primary CD138+ human myeloma cells and myeloma cell lines. Eltrombopag at doses of 0.1 to 100 μM did not enhance proliferation of primary human CD138+ multiple myeloma cells from patients with relapsed disease or myeloma cell lines when used alone or in combination with erythropoietin (EPO) and granulocyte colony-stimulating factor (G-CSF) and did not alter cell viability nor apoptosis of human myeloma cells exposed to bortezomib and lenalidomide. Eltrombopag stimulated megakaryopoiesis in human CD34+ cells from normal individuals and from patients with relapsed multiple myeloma via activation of Akt signaling pathways. These results provide proof-of-principle supporting the design of future clinical studies examining eltrombopag for the treatment of thrombocytopenia in patients with advanced multiple myeloma.
Sigma-Aldrich
Stem Cell Factor human, SCF, recombinant, expressed in HEK 293 cells, suitable for cell culture
Sigma-Aldrich
SCF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Stem Cell Factor human, SCF, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Supelco
Aphidicolin, analytical standard
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)