MilliporeSigma
Expression of nestin after renal transplantation in the rat.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica (2014-04-05)
Michael Skwirba, Anna Zakrzewicz, Srebrena Atanasova, Sigrid Wilker, Gabriele Fuchs-Moll, Dieter Müller, Winfried Padberg, Veronika Grau
ABSTRACT
Chronic allograft injury (CAI) limits the long-term success of renal transplantation. Nestin is a marker of progenitor cells, which probably contribute to its pathogenesis. We hypothesize that nestin is induced by ischemia/reperfusion injury and acute rejection, main risk factors for CAI. Syngeneic renal transplantation was performed in Lewis rats and allogeneic transplantation in the Fischer 344 to Lewis strain combination, which results in reversible acute rejection and in CAI in the long-run. The Dark Agouti to Lewis rat strain combination was used to study fatal acute rejection. In untreated kidneys, nestin immunoreactivity was detected in glomeruli and in very few interstitial or microvascular cells. Syngeneic transplantation induced nestin expression within 4 days, which decreased until day 9 and returned to control levels on day 42. Nestin expression was strong during acute rejection and still detected during the pathogenesis of CAI on day 42. Nestin-positive cells were identified as endothelial cells and interstitial fibroblast-like cells co-expressing alpha-smooth muscle actin. A sub-population of them expressed proliferating cell nuclear antigen. In conclusion, nestin is induced in renal grafts by ischemia/reperfusion injury and acute rejection. It is expressed by proliferating myofibroblasts and endothelial cells and probably contributes to the pathogenesis of CAI.
Product Description
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, BioXtra