Reproductive toxicity of lead, cadmium, and phthalate exposure in men.
Environmental science and pollution research international (2014-05-13)
Niraj Pant, G Kumar, A D Upadhyay, D K Patel, Y K Gupta, P K Chaturvedi
ABSTRACT
Environmental toxicants viz lead or cadmium and phthalate esters (di(2-ethylhexyl) phthalate [DEHP], dibutyl phthalate [DBP], and diethyl phthalate [DEP]) widely found in different environmental strata are linked to deteriorating male reproductive health. The objective was to assess the relationships between the seminal lead, cadmium, and phthalate (DEHP, DBP, DEP) concentrations at environmental level and serum hormone levels and semen quality in non-occupationally exposed men and specify the effect of individual and combined exposure of toxicants on semen quality. A study of 60 male partners of couples attending the Andrology Laboratory of the Reproductive Biology Department, All India Institute of Medical Sciences (AIIMS), New Delhi, India for semen analysis to assess their inability to achieve a pregnancy was selected for the study. The results of univariate and stepwise multiple regression analysis in the unadjusted model showed a significant correlation between lead or cadmium and phthalates DEHP/DBP/DEP and sperm motility, sperm concentration, and DNA damage. After adjusting for potential confounders, an association with lead or DEHP was only observed. The present data shows that lead (Pb) or cadmium (Cd) or phthalates might independently contribute to decline in semen quality and induce DNA damage. Phthalates might influence reproductive hormone testosterone. These findings are significant in light of the fact that men are exposed to a volley of chemicals; however, due to the small sample size, our finding needs to be confirmed in a larger population.
Product Description
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Supelco
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Supelco
Bis(2-ethylhexyl) phthalate, Selectophore
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Supelco
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Plastic additive 01, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, red, fuming, HNO3 >90 %
Sigma-Aldrich
Nitric acid, ACS reagent, 70%