  • Search Results
  • Direct activating effects of adrenocorticotropic hormone (ACTH) on brown adipose tissue are attenuated by corticosterone.
Direct activating effects of adrenocorticotropic hormone (ACTH) on brown adipose tissue are attenuated by corticosterone.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2014-08-03)
Johanna C van den Beukel, Aldo Grefhorst, Carmelo Quarta, Jacobie Steenbergen, Pier G Mastroberardino, Marc Lombès, Patric J Delhanty, Roberta Mazza, Uberto Pagotto, Aart Jan van der Lely, Axel P N Themmen
ABSTRACT
Brown adipose tissue (BAT) and brown-like cells in white adipose tissue (WAT) can dissipate energy through thermogenesis, a process mediated by uncoupling protein 1 (UCP1). We investigated whether stress hormones ACTH and corticosterone contribute to BAT activation and browning of WAT. ACTH and corticosterone were studied in male mice exposed to 4 or 23°C for 24 h. Direct effects were studied in T37i mouse brown adipocytes and primary cultured murine BAT and inguinal WAT (iWAT) cells. In vivo effects were studied using (18)F-deoxyglucose positron emission tomography. Cold exposure doubled serum ACTH concentrations (P=0.03) and fecal corticosterone excretion (P=0.008). In T37i cells, ACTH dose-dependently increased Ucp1 mRNA (EC50=1.8 nM) but also induced Ucp1 protein content 88% (P=0.02), glycerol release 32% (P=0.03) and uncoupled respiration 40% (P=0.003). In cultured BAT and iWAT, ACTH elevated Ucp1 mRNA by 3-fold (P=0.03) and 3.7-fold (P=0.01), respectively. In T37i cells, corticosterone prevented induction of Ucp1 mRNA and Ucp1 protein by both ACTH and norepinephrine in a glucocorticoid receptor (GR)-dependent fashion. ACTH and GR antagonist RU486 independently doubled BAT (18)F-deoxyglucose uptake (P=0.0003 and P=0.004, respectively) in vivo. Our results show that ACTH activates BAT and browning of WAT while corticosterone counteracts this.
Sigma-Aldrich
Corticosterone, ≥92%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Supelco
Corticosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard