  • Search Results
  • Viral suppressors of the RIG-I-mediated interferon response are pre-packaged in influenza virions.
Viral suppressors of the RIG-I-mediated interferon response are pre-packaged in influenza virions.
Nature communications (2014-12-10)
Swantje Liedmann, Eike R Hrincius, Cliff Guy, Darisuren Anhlan, Rüdiger Dierkes, Robert Carter, Gang Wu, Peter Staeheli, Douglas R Green, Thorsten Wolff, Jonathan A McCullers, Stephan Ludwig, Christina Ehrhardt
ABSTRACT
The type I interferon (IFN) response represents the first line of defence to invading pathogens. Internalized viral ribonucleoproteins (vRNPs) of negative-strand RNA viruses induce an early IFN response by interacting with retinoic acid inducible gene I (RIG-I) and its recruitment to mitochondria. Here we employ three-dimensional stochastic optical reconstruction microscopy (STORM) to visualize incoming influenza A virus (IAV) vRNPs as helical-like structures associated with mitochondria. Unexpectedly, an early IFN induction in response to vRNPs is not detected. A distinct amino-acid motif in the viral polymerases, PB1/PA, suppresses early IFN induction. Mutation of this motif leads to reduced pathogenicity in vivo, whereas restoration increases it. Evolutionary dynamics in these sequences suggest that completion of the motif, combined with viral reassortment can contribute to pandemic risks. In summary, inhibition of the immediate anti-viral response is 'pre-packaged' in IAV in the sequences of vRNP-associated polymerase proteins.
Product Description
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, 0.2 μm filtered
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Xylazine, ≥99%
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Tris(tert-butoxy)silanol, packaged for use in deposition systems
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Isoflurane, European Pharmacopoeia (EP) Reference Standard