MilliporeSigma
  • Single adult kidney stem/progenitor cells reconstitute three-dimensional nephron structures in vitro.
Single adult kidney stem/progenitor cells reconstitute three-dimensional nephron structures in vitro.
Stem cells (Dayton, Ohio) (2014-11-26)
Shinji Kitamura, Hiroyuki Sakurai, Hirofumi Makino
ABSTRACT
The kidneys are formed during development from two distinct primordial tissues, the metanephric mesenchyme and the ureteric bud. The metanephric mesenchyme develops into the kidney nephron, the minimal functional unit of the kidney. A nephron consists of several segments and regulates water, electrolyte, and acid-base homeostasis in addition to secreting certain hormones. It has been predicted that the kidney will be among the last organs successfully regenerated in vitro due to its complex structure and multiple functions. Here, we show that adult kidney stem/progenitor cells (KS cells), derived from the S3 segment of adult rat kidney nephrons, can reconstitute a three-dimensional kidney-like structure in vitro. Kidney-like structures were formed when a cluster of KS cells was suspended in an extracellular matrix gel and cultured in the presence of several growth factors. Morphological analyses revealed that these kidney-like structures contained every substructure of the kidney, including glomeruli, proximal tubules, the loop of Henle, distal tubules, and collecting ducts, but no vasculature. Our results demonstrate that a cluster of tissue stem/progenitor cells has the ability to reconstitute the minimum unit of its organ of origin by differentiating into specialized cells in the correct location. This process differs from embryonic kidney development, which requires the mutual induction of two different populations of progenitors, metanephric mesenchymal cells and ureteric bud cells.
Product Description
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pirarubicin, ≥95% (HPLC)
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Selenous acid, 98%
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Selenous acid, 99.999% trace metals basis
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%