  • Search Results
  • Phospholipid profile of amniotic fluid in ovine model of congenital diaphragmatic hernia (CDH): the effect of fetal tracheal occlusion.
Phospholipid profile of amniotic fluid in ovine model of congenital diaphragmatic hernia (CDH): the effect of fetal tracheal occlusion.
Journal of proteome research (2015-01-28)
Maria Chiara Mimmi, Maurizio Ballico, Francesco Amoroso, Valeria Calcaterra, Mario Marotta, Jose Luis Peiro, Gloria Pelizzo
ABSTRACT
Fetal endoscopic tracheal occlusion has been proposed as a prenatal intervention to ameliorate congenital diaphragmatic hernia (CDH) prognosis. Tracheal occlusion (TO) prevents pulmonary fluid egress, leading to tissue expansion, reversal of lung hypoplasia, and potential maturation. Fetal lung maturity strongly correlates with amniotic fluid (AF) phospholipidic composition. In this preliminary study, we characterized the AF phospholipidic profile in CDH-induced, TO-treated, and healthy fetal lambs to define the prenatal treatment benefits of TO on lung maturity. CDH induction was performed at 70 days of gestation, TO was carried out at 102 days of gestation, and caesarean section was carried out at 136 days of gestation. AF samples, taken at 102-136 days of gestation, were evaluated using mass spectrometry. The analysis focused on phosphatidylcholines (PCs) and sphingomyelins (SMs). The most abundant phosphatidylcholine species retrieved in healthy AF was POPC [PC(18:1/16:0)], while the level of DPPC [PC(16:0/16:0)] was extremely low at both gestational ages. CDH induction caused a decrease in POPC and many other PCs. A substantial return of some PCs, in particular POPC, PC(34:2) and PC(18:0/16:0), to a more physiological level was prompted by TO. SMs were unaltered. The AF phospholipidic profile could provide prenatal prognostic markers of CDH and possible indices of lung maturation after fetal treatment.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Methanol, NMR reference standard