  • Search Results
  • A sensitive quantification of the peptide apidaecin 1 isoforms in single bee tissues using a weak cation exchange pre-separation and nanocapillary liquid chromatography coupled with mass spectrometry.
A sensitive quantification of the peptide apidaecin 1 isoforms in single bee tissues using a weak cation exchange pre-separation and nanocapillary liquid chromatography coupled with mass spectrometry.
Journal of chromatography. A (2014-12-02)
Jiří Danihlík, Marek Šebela, Marek Petřivalský, René Lenobel
ABSTRACT
Apidaecins represent an important group of antimicrobial peptides occurring in honey bee hemolymph, where they play an important role as key components of humoral immunity. The present study demonstrates the development of a highly sensitive assay for apidaecin 1 isoforms quantification in the hemolymph or body parts from honey bee individuals. The analytical protocol comprises apidaecins 1 purification and enrichment steps by weak cation-exchange chromatography (WCX) in laboratory-made WCX-Tip microcolumns combined with a desalting step on a reversed-phase sorbent (C8) carried in StageTips. Apidaecin-enriched fraction was analyzed by a reversed-phase based nanoliquid chromatography (C4) separation coupled with high-resolution mass spectrometry. The method performance was validated in its specificity, linearity (0-5pmol), recovery (∼45%), precision (<10% at 0.1pmol), limit of detection (∼50fmol), limit of quantification (0.1pmol) and sample stability. The method was successfully applied to analyze the content of apidaecin 1 isoforms in the following samples: hemolymph - 13.0ng/μL (95% confidence interval of 7.5-18.6ng/μL), thoraxes - 36.2ng/unit (95% CI of 18.9-53.6ng/unit) and heads - 12.9ng/unit (95% CI of 9.1-16.7ng/unit). Freshly emerged bees had apidaecin 1 isoforms levels below the limit of detection. Thus it was possible to use them as a competitive matrix for calibration standards to prevent losses of highly basic apidaecins. This new protocol for apidaecin 1 isoforms quantification represents a promising tool to study the role of apidaecins in honey bee immunity and can be considered as a proof-of-concept for the development of sensitive quantification methods for basic antimicrobial peptides in various organisms.
Product Description
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Supelco
2-Propanol, analytical standard
Supelco
Acetonitrile, analytical standard
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)