  • Search Results
  • Development and validation of a LC-MS/MS method for the quantitation of lumefantrine in mouse whole blood and plasma.
Development and validation of a LC-MS/MS method for the quantitation of lumefantrine in mouse whole blood and plasma.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-02-04)
Katya Govender, Liezl Gibhard, Lissinda Du Plessis, Lubbe Wiesner
ABSTRACT
A liquid chromatography-tandem mass spectrometry (LC-MS/MS) method was developed and validated for the quantitation of the antimalarial drug, lumefantrine (LF), in mouse whole blood and plasma. The analyte was extracted using a protein precipitation method followed by chromatographic separation on a Phenomenex Luna, PFP (50mm×2.0mm, 5μm) analytical column with a mobile phase consisting of acetonitrile and 0.1% formic acid (formic acid:water, 1:1000, v/v) at a ratio of 3:7 (v/v), delivered at a constant flow rate of 0.5ml/min. Stable isotope labeled lumefantrine (D9-LF) was used as the internal standard. Multiple reaction monitoring was performed using the transitions m/z 530.1→m/z 347.9 and m/z 539.1→m/z 347.9 for the quantification of LF and D9-LF, respectively. Calibration curves were constructed over the concentration range 15.6-4000ng/ml. The mean intra- and inter-assay accuracy values for the analysis of LF in WB was 103% (%CV=5.5) and 99.5% (%CV=5.5), respectively. The mean intra- and inter-assay accuracy values for the analysis of LF in plasma was 93.7% (%CV=3.5) and 93.9% (%CV=5.5), respectively. No significant matrix effect was observed during the method validation. The validated method was applied to an absorption study in mice, to determine and compare LF concentrations in whole blood and plasma samples. Results of the statistical analysis using a linear mixed effects growth curve model concluded that there was no significant difference (p-value=0.668) between WB and plasma LF concentrations. This method utilizes a small sample volume of 20μl, facilitating low blood collection volumes and a short chromatographic run time of 3min which allows for high sample throughput analysis.
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%