  • Search Results
  • Role of electron transport chain of chloroplasts in oxidative burst of interaction between Erwinia amylovora and host cells.
Role of electron transport chain of chloroplasts in oxidative burst of interaction between Erwinia amylovora and host cells.
Photosynthesis research (2015-03-31)
Hamid Abdollahi, Zahra Ghahremani, Kobra Erfaninia, Rahim Mehrabi
ABSTRACT
Erwinia amylovora is a necrogenic bacterium, causing the fire blight disease on many rosaceous plants. Triggering oxidative burst by E. amylovora is a key response by which host plants try to restrain pathogen spread. Electron transport chain (ETC) of chloroplasts is known as an inducible source of reactive oxygen species generation in various stresses. This research was performed to assess the role of this ETC in E. amylovora-host interaction using several inhibitors of this chain in susceptible and resistant apple and pear genotypes. All ETC inhibitors delayed appearance of disease necrosis, but the effects of methyl viologen, glutaraldehyde, and DCMU were more significant. In the absence of inhibitors, resistant genotypes showed an earlier and severe H2O2 generation and early suppression of redox dependent, psbA gene. The effects of inhibitors were corresponding to the redox potential of ETC inhibitory sites. In addition, delayed necrosis appearance was associated with the decreased disease severity and delayed H2O2 generation. These results provide evidences for the involvement of this ETC in host oxidative burst and suggest that chloroplast ETC has significant role in E. amylovora-host interaction.
Product Description
Supelco
Uracil, Pharmaceutical Secondary Standard; Certified Reference Material
Fluorouracil impurity C, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Uracil, BioReagent, suitable for cell culture
Sigma-Aldrich
Uracil, ≥99.0%
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Diuron, ≥98%
Supelco
Diuron, PESTANAL®, analytical standard
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Methyl viologen dichloride hydrate, 98%
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O