MilliporeSigma
  • Rapid access to glycopeptide antibiotic precursor peptides coupled with cytochrome P450-mediated catalysis: towards a biomimetic synthesis of glycopeptide antibiotics.
Rapid access to glycopeptide antibiotic precursor peptides coupled with cytochrome P450-mediated catalysis: towards a biomimetic synthesis of glycopeptide antibiotics.
Organic & biomolecular chemistry (2014-12-17)
Clara Brieke, Veronika Kratzig, Kristina Haslinger, Andreas Winkler, Max J Cryle
ABSTRACT
Understanding the mechanisms underpinning glycopeptide antibiotic biosynthesis is key to the future ability to reinvent these compounds. For effective in vitro characterization of the crucial later steps of the biosynthesis, facile access to a wide range of substrate peptides as their Coenzyme A (CoA) conjugates is essential. Here we report the development of a rapid route to glycopeptide precursor CoA conjugates that affords both high yields and excellent purities. This synthesis route is applicable to the synthesis of peptide CoA-conjugates containing racemization-prone arylglycine residues: such residues are hallmarks of non-ribosomal peptide synthesis and have previously been inaccessible to peptide synthesis using Fmoc-type chemistry. We have applied this route to generate glycopeptide precursor peptides in their carrier protein-bound form as substrates to explore the specificity of the first oxygenase enzyme from vancomycin biosynthesis (OxyBvan). Our results indicate that OxyBvan is a highly promiscuous catalyst for phenolic coupling of diverse glycopeptide precursors that accepts multiple carrier protein substrates, even on carrier protein domains from alternate glycopeptide biosynthetic machineries. These results represent the first important steps in the development of an in vitro biomimetic synthesis of modified glycopeptide aglycones.
Sigma-Aldrich
Nitrogen, ≥99.998%
Supelco
trans-Sinapic acid, analytical standard
Sigma-Aldrich
Nitrilotriacetic acid disodium salt, Sigma Grade, ≥99%
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Supelco
Sinapic acid, matrix substance for MALDI-MS, ≥99.5%, Ultra pure
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Imidazole, European Pharmacopoeia (EP) Reference Standard
USP
Imidazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Ondansetron impurity E, European Pharmacopoeia (EP) Reference Standard
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Nitrilotriacetic acid trisodium salt, Sigma Grade, ≥98%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)