  • Search Results
  • Serum and adipose tissue amino acid homeostasis in the metabolically healthy obese.
Serum and adipose tissue amino acid homeostasis in the metabolically healthy obese.
Journal of proteome research (2014-06-17)
Flavia Badoud, Karen P Lam, Alicia DiBattista, Maude Perreault, Michael A Zulyniak, Bradley Cattrysse, Susan Stephenson, Philip Britz-McKibbin, David M Mutch
ABSTRACT
A subgroup of obese individuals, referred to as metabolically healthy obese (MHO), have preserved insulin sensitivity and a normal lipid profile despite being obese. The molecular basis for this improved cardiometabolic profile remains unclear. Our objective was to integrate metabolite and gene expression profiling to elucidate the molecular distinctions between MHO and metabolically unhealthy obese (MUO) phenotypes. A subset of individuals were selected from the Diabetes Risk Assessment study and classified into three groups using anthropometric and clinical measurements: lean healthy (LH), MHO, and MUO. Serum metabolites were profiled using gas chromatography coupled to mass spectrometry. Multivariate data analysis uncovered metabolites that differed between groups, and these were subsequently validated by capillary electrophoresis coupled to mass spectrometry. Subcutaneous adipose tissue (SAT) gene expression profiling using microarrays was performed in parallel. Amino acids were the most relevant class of metabolites distinguishing MHO from MUO individuals. Serum levels of glutamic acid, valine, and isoleucine were positively associated (i.e., LH < MHO < MUO) with homeostasis model assessment-insulin resistance (HOMA-IR) and glycated hemoglobin (HbA1c) values, while leucine was only correlated with HOMA-IR. The glutamine-to-glutamic acid ratio and glycine were inversely correlated (i.e., LH > MHO > MUO) with HbA1c values. Concomitantly, SAT gene expression profiling revealed that genes related to branched-chain amino acid catabolism and the tricarboxylic acid cycle were less down-regulated in MHO individuals compared to MUO individuals. Together, this integrated analysis revealed that MHO individuals have an intermediate amino acid homeostasis compared to LH and MUO individuals.
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chlorotrimethylsilane, for GC derivatization, LiChropur, ≥99.0% (GC)
SAFC
Acetic acid, glacial
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%