  • Search Results
  • Chronic urocortin 2 administration improves cardiac function and ameliorates cardiac remodeling after experimental myocardial infarction.
Chronic urocortin 2 administration improves cardiac function and ameliorates cardiac remodeling after experimental myocardial infarction.
Journal of cardiovascular pharmacology (2015-02-07)
Leigh J Ellmers, Nicola J A Scott, Vicky A Cameron, A Mark Richards, Miriam T Rademaker
ABSTRACT
The impact of chronic urocortin 2 (Ucn2) treatment after myocardial infarction (MI) has not previously been investigated. In this study, we examined the effects of 30-day Ucn2 administration (415 μg·kg·d SC per day) in mice post-MI. Compared with surgical sham + vehicle controls (n = 10), MI + vehicle animals (n = 10) after 30 days demonstrated decreased ejection fraction (75.6 ± 1.2 vs. 43.6% ± 0.8%, P < 0.001) and fractional shortening (38.20 ± 0.83 vs. 18.4% ± 0.54%, P < 0.001) in association with increased heart weight-to-body weight ratio (4.57 ± 0.25 vs. 5.29 ± 0.18, P < 0.01), left ventricular (LV) mass (91 ± 7 vs. 126 ± 8 mg, P < 0.01), LV internal diameters at both systole (1.91 ± 0.14 vs. 3.45 ± 0.09 mm, P < 0.001) and diastole (3.14 ± 0.15 vs. 4.25 ± 0.10 mm, P < 0.001), LV end systolic volumes (0.02 ± 0.01 vs. 0.11 ± 0.01 mL, P < 0.001), and ventricular collagen 1 and β-myosin heavy chain gene expression. Compared with MI + vehicle mice, MI + Ucn2 animals (n = 10) exhibited significantly reduced infarct size (4.00 ± 0.39 vs. 1.83 ± 0.44 mm, P < 0.01), heart weight-to-body weight ratio (4.75 ± 0.19, P = 0.06), LV mass (101 ± 6 mg, P < 0.01), LV internal diameters (systole 2.61 ± 0.09 mm, P < 0.001; diastole 3.78 ± 0.09 mm, P < 0.001), and end systolic volumes (0.14 ± 0.02 mL, P < 0.01) in conjunction with improved ejection fraction (65.2% ± 0.9%, P < 0.001) and fractional shortening (18.4 ± 0.5 vs. 30.5% ± 0.5%, P < 0.001). Ucn2 treatment also decreased collagen 1 and β-myosin heavy chain expression. In conclusion, chronic Ucn2 treatment significantly improves cardiovascular function and attenuates cardiac injury and remodeling in experimental MI.
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Supelco
Chloroform, analytical standard
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis