MilliporeSigma
  • ATF3 functions as a novel tumor suppressor with prognostic significance in esophageal squamous cell carcinoma.
ATF3 functions as a novel tumor suppressor with prognostic significance in esophageal squamous cell carcinoma.
Oncotarget (2014-08-26)
Jian-Jun Xie, Yang-Min Xie, Bo Chen, Feng Pan, Jin-Cheng Guo, Qing Zhao, Jin-Hui Shen, Zhi-Yong Wu, Jian-Yi Wu, Li-Yan Xu, En-Min Li
ABSTRACT
ATF3 was a transcription factor involved in the progression of certain cancers. Here, we sought to explore the expression and biological function of ATF3 in esophageal squamous cell carcinomas (ESCC). The prognostic significance of ATF3 expression was evaluated in 150 ESCC samples and 21 normal squamous cell epithelium tissues. Results showed that ATF3 was down-regulated in ESCC lesions compared with paired non-cancerous tissues and low tumorous ATF3 expression significantly correlated with shorter overall survival (OS) and disease-free survival (DFS). Cox regression analysis confirmed that ATF3 expression was an independent prognostic factor. Experimentally, forced expression of ATF3 led to decreased growth and invasion properties of ESCC cells in vitro and in vivo, whereas knockdown of ATF3 did the opposite. Furthermore, ATF3 upregulated the expression of MDM2 by increasing the nuclear translocation of P53 and formed an ATF3/MDM2/MMP-2 complex that facilitated MMP-2 degradation, which subsequently led to inhibition of cell invasion. Finally, we showed that Cisplatin could restrain the invasion of ESCC cells by inducing the expression of ATF3 via P53 signaling. Combined, our findings highlight a suppressed role for ATF3 in ESCC and targeting ATF3 might be a potential therapeutic strategy.
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
DAPI, for nucleic acid staining
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard