  • Search Results
  • Development of novel sophorolipids with improved cytotoxic activity toward MDA-MB-231 breast cancer cells.
Development of novel sophorolipids with improved cytotoxic activity toward MDA-MB-231 breast cancer cells.
Journal of molecular recognition : JMR (2015-02-04)
Isabel A C Ribeiro, Célia M C Faustino, Patrícia S Guerreiro, Raquel F M Frade, M Rosário Bronze, Matilde F Castro, Maria H L Ribeiro
ABSTRACT
Sophorolipids (SLs) are glycolipid biosurfactants, produced as a mixture of several compounds by some nonpathogenic yeast. In the current study, separation of individual SLs from mixtures with further evaluation of their surface properties and biologic activity on MDA-MB-321 breast cancer cell line were investigated. SLs were biosynthesized by Starmerella bombicola in a culture media supplemented with borage oil. A reverse-phase flash chromatography method with an automated system coupled with a prepacked cartridge was used to separate and purify the main SLs. Compositional analysis of SLs was performed by high-performance liquid chromatography with electrospray ionization mass spectrometry and tandem mass spectrometry. The following diacetylated lactonic SLs were isolated and purified: C18:0, C18:1, C18:2, and C18:3. The critical micelle concentration (CMC) and surface tension at CMC (γCMC ) of the purified SLs showed an increase with the number of double bonds. High cytotoxic effect against MDA-MB-231 cells was observed with C18:0 and C18:1 lactonic SLs. The cytotoxic effects of C18:3 lactonic SL on cancerous cells were for the first time studied. This cytotoxic effect was considerably higher than the promoted by acidic SLs; however, it induced a lower effect than the previously mentioned SLs, C18:0 and C18:1. To our knowledge, for the first time, C18:1 lactonic SL, in selected concentrations, proved to be able to inhibit MDA-MB-231 cell migration without compromising cell viability and to increase intracellular reactive oxygen species.
Product Description
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water
Supelco
Anthrone, analytical standard
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Sodium sulfate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., anhydrous, ≥99.0%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)