MilliporeSigma
  • A validated chiral LC method for enantiomeric separation of nebivolol stereoisomers in bulk drugs and dosage forms on amylose-based stationary phase.
A validated chiral LC method for enantiomeric separation of nebivolol stereoisomers in bulk drugs and dosage forms on amylose-based stationary phase.
Journal of chromatographic science (2013-10-31)
Karri Visweswara Rao, Kesareddy Padmaja Reddy, Pranab Haldar
ABSTRACT
A novel and reproducible isocratic normal phase liquid chromatographic method was developed for the quantitative determination of 10 stereoisomers of Nebivolol in pharmaceutical bulk drugs and dosage forms. The method was developed using an amylose-based chiral stationary phase, Chiralpak AD-3 (250 × 4.6 mm, 3 μm) column with mobile phase containing n-hexane-ethanol-isopropanol-diethanolamine in the ratio 42:45:13:0.1 (v/v/v/v). The eluted compounds were monitored at 280 nm. Ten stereoisomers of Nebivolol were well separated with resolution >2.0 for all pair of components. The developed method was validated as per International Conference on Harmonization (ICH) guidelines with respect to specificity, linearity (R(2) value >0.999), limit of detection, limit of quantification, accuracy (recovery range 95.8-103.2%), precision (relative standard deviation, RSD, <2.5%) and robustness. Nebivolol sample solutions were found to be stable when characterized over a period of 48 h. Forced degradation studies were also performed to demonstrate the stability-indicating power of the developed HPLC method. The method was found to be rugged and robust.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Supelco
Hexane, analytical standard
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)