  • Search Results
  • Identification of ultraviolet B radiation‑induced microRNAs in normal human dermal papilla cells.
Identification of ultraviolet B radiation‑induced microRNAs in normal human dermal papilla cells.
Molecular medicine reports (2014-07-30)
Hwa Jun Cha, Ok-Yeon Kim, Gang Tai Lee, Kwang Sik Lee, Jae Ho Lee, In-Chul Park, Su-Jae Lee, Yu Ri Kim, Kyu Joong Ahn, In-Sook An, Sungkwan An, Seunghee Bae
ABSTRACT
Ultraviolet (UV) radiation impairs intracellular functions by directly damaging DNA and by indirectly generating reactive oxygen species (ROS), which induce cell cycle arrest and apoptosis. UV radiation can also alter gene expression profiles, including those of mRNA and microRNA (miRNA). The effects of UV radiation on cellular functions and gene expression have been widely documented in human skin cells such as keratinocytes, melanocytes and dermal fibroblasts, but the effect it has on other types of skin cell such as dermal papilla cells, which are crucial in the induction of hair follicle growth, remains unknown. In the current study, the effect of UV radiation on physiological changes and miRNA-based expression profiles in normal human dermal papilla cells (nHDPs) was investigated. UVB radiation of ≥50 mJ/cm2 displayed high cytotoxicity and apoptosis in a dose-dependent manner. In addition, ROS generation was exhibited in UVB-irradiated nHDPs. Furthermore, using miRNA microarray analysis, it was demonstrated that the expression profiles of 42 miRNAs in UVB-irradiated nHDPs were significantly altered compared with those in the controls (35 upregulated and 7 downregulated). The biological functions of the differentially expressed miRNAs were studied with gene ontology analysis to identify their putative target mRNAs, and were demonstrated to be involved in cell survival- and death-related functions. Overall, the results of the present study provide evidence that miRNA‑based cellular mechanisms may be involved in the UVB-induced cellular response in nHDPs.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
2′,7′-Dichlorofluorescein diacetate, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard