  • Search Results
  • Potentiometric aptasensing of Listeria monocytogenes using protamine as an indicator.
Potentiometric aptasensing of Listeria monocytogenes using protamine as an indicator.
Analytical chemistry (2014-09-16)
Jiawang Ding, Jiahong Lei, Xia Ma, Jun Gong, Wei Qin
ABSTRACT
Exposure to pathogens in recreational or drinking water is a serious public health concern. It is important to rapidly determine and identify trace levels of pathogens in real environmental samples. We report here on a label-free potentiometric aptasensor for rapid, sensitive, and selective detection of Listeria monocytogenes (LM), a pathogen widely distributed in the environment. An aptamer binds specifically to internalin A, a surface protein present in LM cells. The target-binding event prevents the aptamer from electrostatically interacting with protamine, which can be sensitively detected using a polycation-sensitive membrane electrode. Using this method, LM can be detected down to 10 CFU mL(-1). Coupled to an online filtration system, the bioassay has been evaluated with spiked coastal seawater samples and shows good recovery and high accuracy. This work demonstrates the possibility of developing potentiometric aptasensors for determination and identification of various bacteria in environmental samples.
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Heptane, biotech. grade, ≥99%
Sigma-Aldrich
Heptane, anhydrous, 99%
Supelco
Heptane, HPLC grade, ≥99%
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Supelco
Heptane, analytical standard
Supelco
Trizma® base, certified reference material for titrimetry, certified by BAM, ≥99.5%
Supelco
Heptane, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Heptane, puriss. p.a., reag. Ph. Eur., ≥99% n-heptane basis (GC)
Sigma-Aldrich
Heptane, ReagentPlus®, 99%
Sigma-Aldrich
Heptane, suitable for HPLC, ≥99%
Sigma-Aldrich
Heptane, suitable for HPLC, ≥96%
Sigma-Aldrich
Heptane, HPLC Plus, for HPLC, GC, and residue analysis, 99%
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Supelco
2-Nitrophenyl octyl ether, Selectophore, ≥99.0%
Sigma-Aldrich
2-Nitrophenyl octyl ether, 99%
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material
SAFC
Tromethamine
Trometamol, European Pharmacopoeia (EP) Reference Standard
USP
Tromethamine, United States Pharmacopeia (USP) Reference Standard