  • Search Results
  • Chitosan nanoparticles for siRNA delivery: optimization of processing/formulation parameters.
Chitosan nanoparticles for siRNA delivery: optimization of processing/formulation parameters.
Nucleic acid therapeutics (2014-10-02)
Elina Esmaeilzadeh Gharehdaghi, Amir Amani, Mohammad Reza Khoshayand, Mehdi Banan, Elika Esmaeilzadeh Gharehdaghi, Mohammad Ali Amini, Mohammad Ali Faramarzi
ABSTRACT
Chitosan nanoparticles were prepared using ultrasonication methodology at specific amplitudes and times of sonication. Subsequently, small interfering RNA (siRNA) was added to the solution at predetermined values of nitrogen to phosphorous ratio (N/P), and stirring time. Employing response surfaces generated from a statistical model, the effect of sonication time and amplitude, stirring time, and N/P ratio was studied on the particle size, polydispersity, and loading efficiency of prepared siRNA/chitosan nanoparticles. It was found that to obtain the smallest size, amplitude and time of sonication as well as stirring time should be kept at ∼45%, 165 seconds, and 50 minutes, respectively. Minimum polydispersity values were also obtained at similar values of sonication time/amplitude and stirring time in addition to N/P values of ∼28. Also, the maximum proportion of siRNA loading was observed at approximate values of 300 seconds, 80% and 280 for sonication time, amplitude, and N/P ratio, respectively. The optimum conditions (i.e., to prepare a sample with minimum values of particle size and polydispersity index and maximum values of loading efficiency) were determined as 60.6, 30.0 (seconds), 28.0, and 12.5 (minutes) for amplitude, time of sonication, N/P, and stirring time, respectively. In this scrutiny, the predicted values of optimum formulation were 456 nm size, 89.6% loading efficiency, and 0.4 polydispersity index.
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Diethyl pyrocarbonate, ≥97% (NMR)
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Diethyl pyrocarbonate, 99% (NT)
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Diethyl pyrocarbonate, 97%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard