MilliporeSigma
  • Brain-derived neurotrophic factor accelerates gut motility in slow-transit constipation.
Brain-derived neurotrophic factor accelerates gut motility in slow-transit constipation.
Acta physiologica (Oxford, England) (2014-08-29)
F Chen, Y Yu, P Wang, Y Dong, T Wang, X Zuo, Y Li
ABSTRACT
Brain-derived neurotrophic factor (BDNF) may play a critical role in gut motility. We aimed to investigate BDNF's physiologic effects on gut motility in slow-transit constipation (STC) and to explore the underlying molecular mechanisms. BDNF expression and alterations of colonic nerve fibre density in STC patients were first investigated. BDNF's effects on gastrointestinal motility of both BDNF(+/-) mice and loperamide-induced constipation mice were then examined in vivo and in vitro. Smooth muscle α-actin (α-SMA) expression, and nerve fibre, neuromuscular junction (NMJ) and smooth muscle cell (SMC) alterations were investigated. Finally, the effects of BDNF-induced TrkB-phospholipase C/inositol trisphosphate (TrkB-PLC/IP3) pathway activation on gut motility were investigated. In STC patients, BDNF expression and nerve fibre density were decreased, and mucosal nerve fibre ultrastructural degenerations were demonstrated. Gut motility was decreased in vivo and in vitro in BDNF(+/-) and constipation mice, with BDNF dose-dependently increasing gut motility. In BDNF(+/-) mice, α-SMA expression and nerve fibre density were decreased, and nerve fibre, NMJ and SMC ultrastructural degenerations were observed. Finally, TrkB-PLC/IP3 pathway antagonists dramatically attenuated BDNF's excitatory effect on gut motility, and exogenous BDNF induced an obvious increase in IP3 expression. BDNF plays an important regulatory role in gut motility in STC. It was mediated by altering the intestinal innervation structure, as well as smooth muscle secondary degeneration through a mechanism involving TrkB-PLC/IP3 pathway activation.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Brain-derived neurotrophic factor human, BDNF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material