  • Search Results
  • Comparative analysis of antioxidant activity and functional components of the ethanol extract of lotus (Nelumbo nucifera) from various growing regions.
Comparative analysis of antioxidant activity and functional components of the ethanol extract of lotus (Nelumbo nucifera) from various growing regions.
Journal of agricultural and food chemistry (2014-06-17)
Xu Zhao, Jian Shen, Kyung Ja Chang, Sung Hoon Kim
ABSTRACT
The variations in antioxidant activity and concentration of functional components in the ethanol extracts of lotus seeds and rhizomes based on the growing region and dryness were investigated. Free radical scavenging activity, total phenolic and flavonoid content, and concentration of several specific flavonoids and alkaloids in the ethanol extracts of lotus were measured. Antioxidant activity and its correlative total phenolic content varied characteristically depending on the growing region and dryness. High-perfomance liquid chromatography analysis showed that the ethanol extracts of lotus seeds from Vietnam (Ho Chi Minh City), raw rhizomes from Korea (Siheung), and dried rhizomes from Japan (Nigata) had the greatest specific flavonoid content. The ethanol extracts of seeds from China (Hubei), raw rhizomes from Japan (Nigata), and dried rhizomes from Korea (Siheung) had the greatest specific alkaloid content. Astragaline, rutin, isoquercetin, nuciferine, dauricine, isoliensinine, and neferine were identified in lotus rhizomes for the first time in this study.
Product Description
(±)-Catechin hydrate, primary reference standard
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Supelco
(+)-Catechin, analytical standard
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Supelco
Phenol, PESTANAL®, analytical standard
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Supelco
Gallic acid, certified reference material, TraceCERT®
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, natural, 97%, FG
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology