  • Search Results
  • Anti-apoptotic potency of TNFR:Fc gene in ischemia/ reperfusion-induced myocardial cell injury.
Anti-apoptotic potency of TNFR:Fc gene in ischemia/ reperfusion-induced myocardial cell injury.
Inflammation (2014-07-13)
Jun Guo, Dong Zheng, Hai-Rui Li, Ai-Dong Zhang, Zi-Cheng Li
ABSTRACT
The aim of the study was to investigate the anti-apoptotic potency of TNFR:Fc gene in ischemia/reperfusion-induced myocardial cell injury and hypoxia/reoxygenation-induced H9c2 rat cardiomyocytes injury. Rats were randomly divided into the following groups (n=8): (1) sham operation group; (2) ischemia-reperfusion (I/R) rats treated with rAAV-EGFP; (3) I/R rats treated with rAAV-TNFR:Fc group. rAAV-EGFP or rAAV-TNFR:Fc was injected intra-myocardial at four sites on the anterior and posterior walls of left ventricle immediately after the construction of I/R-induced AMI model in rats. The effects of TNFR:Fc on apoptosis and cardiacfunction were observed after 72 h of coronary reperfusion. In the in vitro study, apoptosis was analyzed in H9c2 rat cardiomyocytes treated either with nomoxia alone, or hypoxia/reoxygenation in the presence of rAAV-GFP or rAAV-TNFR:Fc. We found that (1) TNFR:Fc gene improved cardiac function (EF, LVESP, LVEDP and dp/dt max) post I/R-induced AMI; (2) TNFR:Fc gene inhibited I/R-induced apoptosis and attenuated the level of TNF-α in serum and cardiac tissue; (3) TNFR:Fc gene prevented apoptosis in hypoxia/reoxygenation-induced H9c2 rat cardiomyocytes associated with inhibition of caspase-3 activation and normalization of ratio of the Bcl-2/Bax. We concluded that TNFR:Fc gene transfection has anti-apoptotic potency in ischemia/reperfusion-induced myocardial cell injury.
Sigma-Aldrich
Titanium(IV) chloride, ≥99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) chloride, ReagentPlus®, 99.9% trace metals basis
Sigma-Aldrich
Titanium(IV) chloride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Titanium(IV) chloride solution, 1.0 M in toluene
Sigma-Aldrich
Titanium(IV) chloride solution, 0.09 M in 20% HCl
Sigma-Aldrich
2,3,5-Triphenyltetrazolium chloride, ≥98.0% (HPLC)
Sigma-Aldrich
Titanium tetrachloride, packaged for use in deposition systems