MilliporeSigma
Loss of plexin-B3 in hepatocellular carcinoma.
Experimental and therapeutic medicine (2015-03-18)
Yuwu Liu, Chang Wu, Ying Wang, Sailan Wen, Junpu Wang, Zhihong Chen, Qiongqiong He, Deyun Feng
ABSTRACT
Plexins are the primary receptors of semaphorins, and participate in the majority of intracellular pathways triggered by semaphorins, including the regulation of cell adhesion and the motility of numerous cell types. Recently, several studies have reported that plexins can significantly affect different aspects of cancer cell biology, and the aberrant expression of plexins has been observed in a wide variety of tumor types. However, the expression and role of plexin-B3 in hepatocellular carcinoma (HCC) is yet to be investigated. In the present study, plexin-B3 expression was measured in 14 paired HCC samples and the corresponding adjacent non-cancerous tissue by quantitative polymerase chain reaction and western blot analysis. The results indicated that the mRNA and protein expression levels of plexin-B3 were downregulated in HCC samples when compared with the corresponding adjacent non-cancerous tissue. In order to elucidate the correlation between clinicopathological data and the expression of plexin-B3 in patients with HCC, 84 HCC archived specimens were analyzed by immunohistochemistry (IHC). The IHC results revealed that the protein expression level of plexin-B3 was lower in the HCC samples compared with the corresponding adjacent non-cancerous tissue, and plexin-B3 underexpression was correlated with the patient gender and tumor size. In conclusion, these results indicated that loss of plexin-B3 in HCC may be of predictive value for the occurrence and progression of HCC. Thus, plexin-B3 may be a promising biomarker for the diagnosis and treatment of tumors in the future.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
SAFC
Sodium deoxycholate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)