  • Search Results
  • Recombinant immunoglobulin A specific for influenza A virus hemagglutinin: production, functional analysis, and formation of secretory immunoglobulin A.
Recombinant immunoglobulin A specific for influenza A virus hemagglutinin: production, functional analysis, and formation of secretory immunoglobulin A.
Viral immunology (2015-02-07)
Kentaro Shoji, Tadanobu Takahashi, Kohta Kurohane, Koki Iwata, Takeshi Matsuoka, Shogo Tsuruta, Takatomo Sugino, Masaki Miyake, Takashi Suzuki, Yasuyuki Imai
ABSTRACT
Secretory immunoglobulin (Ig) A (SIgA), comprised of dimeric IgA and secretory component (SC), is believed to provide a defense mechanism on the mucosal surface. Influenza A virus (IAV) hemagglutinin (HA)-specific SIgA is thought to play an important role in the prevention of IAV infection. However, the topical application of preformed IAV-specific SIgA has not been shown to prevent IAV infection. This is due to the difficulty in the production of antigen-specific IgA monoclonal antibodies (mAbs) and monoclonal SIgA. Here, a recombinant hybrid IgA (HIgA) was established that utilizes variable regions of an HA-specific mouse IgG mAb and the heavy chain constant region of a mouse IgA mAb. We expressed the dimeric HIgA in Chinese hamster ovary-K1 (CHO-K1) cells. When in vitro IAV infection of Madin-Darby canine kidney (MDCK) cells was tested, 10 times lower concentrations of HIgA were able to inhibit it as compared with an HA-specific IgG with the same variable regions. A functional hybrid secretory IgA (HSIgA) was also produced through incubation of the dimeric HIgA with recombinant mouse SC in vitro. It was demonstrated that HSIgA could be separated from the dimeric HIgA on size exclusion chromatography. This study provides a basic strategy for investigating the role of SIgA upon IAV infection on the mucosal surface.
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
4-Chloro-1-naphthol, crystalline
Sigma-Aldrich
4-Chloro-1-naphthol, tablet
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
SAFC
HEPES
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%