  • Search Results
  • Identification of microRNA-135b in stool as a potential noninvasive biomarker for colorectal cancer and adenoma.
Identification of microRNA-135b in stool as a potential noninvasive biomarker for colorectal cancer and adenoma.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-04-03)
Chung Wah Wu, Siew Chien Ng, Yujuan Dong, Linwei Tian, Simon Siu Man Ng, Wing Wa Leung, Wai Tak Law, Tung On Yau, Francis Ka Leung Chan, Joseph Jao Yiu Sung, Jun Yu
ABSTRACT
Detecting microRNA (miRNA) in stool is a novel approach for colorectal cancer (CRC) screening. This study aimed to identify stool-based miRNA as noninvasive biomarkers for detection of CRC and adenoma. A miRNA expression array covering 667 human miRNAs was performed on five pairs of CRC and two pairs of advanced adenoma tissues. The most upregulated miRNAs were validated in 40 pairs of CRC tissues, 16 pairs of advanced adenoma tissues, and 424 stool samples, including 104 CRCs, 169 adenomas, 42 inflammatory bowel diseases (IBD), and 109 healthy controls. miRNA levels were followed-up after removal of lesions. In an array analysis, miR-31 and miR-135b were the most upregulated miRNAs in CRC and advanced adenoma as compared with their adjacent normal tissues (>13-fold increase). In stool samples, level of miR-135b was significantly higher in subjects with CRC (P < 0.0001) or adenomas (P < 0.0001), but not in patients with IBD compared with controls. miR-135b showed a significant increasing trend across the adenoma to cancer sequence (P < 0.0001). Levels of miR-31 were not significantly different among groups. The sensitivity of stool mR-135b was 78% for CRC, 73% for advanced adenoma, and 65% for any adenoma, respectively, with a specificity of 68%. No significant difference in the miR-135b level was found between proximal and distal colorectal lesions. Stool miR-135b dropped significantly upon removal of CRC or advanced adenoma (P < 0.0001). Stool-based miR-135b can be used as a noninvasive biomarker for the detection of CRC and advanced adenoma. Clin Cancer Res; 20(11); 2994-3002. ©2014 AACR.
Product Description
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, analytical standard
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material