  • Search Results
  • Detection of peritoneal dissemination with near-infrared fluorescence laparoscopic imaging using a liposomal formulation of a synthesized indocyanine green liposomal derivative.
Detection of peritoneal dissemination with near-infrared fluorescence laparoscopic imaging using a liposomal formulation of a synthesized indocyanine green liposomal derivative.
Anticancer research (2015-03-10)
Isamu Hoshino, Tetsuro Maruyama, Hiromichi Fujito, Yutaka Tamura, Akiko Suganami, Hideki Hayashi, Taro Toyota, Yasunori Akutsu, Kentaro Murakami, Yuka Isozaki, Naoki Akanuma, Nobuyoshi Takeshita, Takeshi Toyozumi, Aki Komatsu, Hisahiro Matsubara
ABSTRACT
Although conventional staging laparoscopy (SL) has improved the diagnostic accuracy of peritoneal dissemination, novel technology is needed to increase the sensitivity of SL. We herein describe a new imaging method employing near-infrared (NIR) fluorescence imaging using a liposomal synthesized indocyanine green (ICG) liposomal derivative, LP-ICG-C18. LP-ICG-C18 is a NIR-photoactivating probe in which an ICG fluorophore is covalently conjugated with a phospholipid moiety. Nude mice were intraperitoneally injected with gastric cancer cells. Twelve days later, the mice were given intravenous injections of LP-ICG-C18 at a dose of 0.15 mg/kg. A NIR imaging system was used to identify the disseminated tumors. The disseminated nodules in mice were detected without any difficulties. Disseminated tumor nodules were collected from mice with or without injections of liposomal formulation and were transferred into the swine peritoneal cavity. The nodules in the swine peritoneal cavity were clearly and promptly defined by the NIR imaging system. NIR-fluorescing liposomal probes can effectively target peritoneal disseminated tumors and can be easily detected by a NIR imaging system. These results warrant future clinical trials of our imaging system and may contribute to a more precise diagnosis and therapeutic approach for gastric cancer patients.
Sigma-Aldrich
Cardiogreen, for microscopy
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Cardiogreen, polymethine dye
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)