MilliporeSigma
  • Automated quantitative analysis of tissue microarray of 443 patients with colorectal adenocarcinoma: low expression of Bcl-2 predicts poor survival.
Automated quantitative analysis of tissue microarray of 443 patients with colorectal adenocarcinoma: low expression of Bcl-2 predicts poor survival.
Journal of the American College of Surgeons (2014-08-16)
Allen D Nicholson, Xiaojia Guo, Catherine A W Sullivan, Charles H Cha
ABSTRACT
Bcl-2 has been implicated in the development and progression of a number of cancers including colorectal cancer. Reports of Bcl-2 expression in colorectal cancer and patient outcomes have been inconsistent due to small cohorts and semi-quantitative grading methods. We used a high throughput tissue microarray system (automated quantitative analysis [AQUA]), analyzing colorectal adenocarcinoma samples from 443 patients resected during the period of 1967 to 1986. This system uses fully quantitative, automated fluorescent microscopy to accurately assess Bcl-2 expression in colorectal cancer samples. Clinicopathologic variables were collected prospectively and were assessed using log-rank tests and Cox proportional hazards models. At a median follow-up of 54 months, the 5- and 10-year disease-specific survivals for all patients were 59.2% and 52.1%, respectively. Loss of Bcl-2 expression was seen in 70.4% of tumors and was associated with a decreased 5-year disease-specific survival (55.8% vs 75.6%, p = 0.001 and relative risk [RR] 1.8) and decreased 5-year overall survival (45.8% vs 56.5%, p = 0.046 and RR 1.2). On univariate analysis, T stage, N stage, and loss of Bcl-2 expression predicted poor disease-specific survival. On multivariate analysis, Bcl-2 expression was an independent prognostic factor for disease-specific survival (p = 0.034). Our results indicate that loss of Bcl-2 expression in colorectal cancer is associated with decreased disease-specific and overall survival. This finding may help identify a subset of patients with a more aggressive phenotype and guide adjuvant chemotherapy choices.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Tris(tert-butoxy)silanol, packaged for use in deposition systems
Sigma-Aldrich
2-Phenylindole, technical grade, 95%