  • Search Results
  • Profiling of Cxcl12 receptors, Cxcr4 and Cxcr7 in murine testis development and a spermatogenic depletion model indicates a role for Cxcr7 in controlling Cxcl12 activity.
Profiling of Cxcl12 receptors, Cxcr4 and Cxcr7 in murine testis development and a spermatogenic depletion model indicates a role for Cxcr7 in controlling Cxcl12 activity.
PloS one (2014-12-03)
Birgit Westernströer, Nicole Terwort, Jens Ehmcke, Joachim Wistuba, Stefan Schlatt, Nina Neuhaus
ABSTRACT
In mice the chemokine Cxcl12 and its receptor Cxcr4 participate in maintenance of the spermatogonial population during postnatal development. More complexity arises since Cxcl12 also binds to the non-classical/atypical chemokine receptor Cxcr7. We explored the expression pattern of Cxcl12, Cxcr4 and Cxcr7 during postnatal development in mouse testes and investigated the response of Cxcl12, Cxcr4, Cxcr7 and SSC-niche associated factors to busulfan-induced germ cell depletion and subsequent recovery by RNA expression analysis and localization of the proteins. In neonatal testes transcript levels of Cxcl12, Cxcr4 and Cxcr7 were relatively low and protein expression of Cxcr7 was restricted to gonocytes and spermatogonia. During development, RNA expression of Cxcl12 remained stable but that of Cxcr4 and Cxcr7 increased. Cxcr7 was expressed in germ cells located at the basement membrane of the seminiferous tubules. In adult testes, transcript levels of Cxcl12 were highest while the localization of Cxcr7 did not change. Following germ cell depletion, a significantly increased expression of Cxcl12 and a decreased expression of Cxcr7 were observed. Germ cells repopulating the seminiferous tubules were immunopositive for Cxcr7. We conclude that Cxcr7 expression to be restricted to premeiotic germ cells throughout postnatal testicular development and during testicular recovery. Hence, the spermatogonial population may not only be simply controlled by interaction of Cxcl12 with Cxcr4 but may also involve Cxcr7 as an important player.
Product Description
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Busulfan, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Streptavidin−Peroxidase from Streptomyces avidinii, lyophilized powder
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
Busulfan, analytical standard, for drug analysis
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Xylazine, ≥99%
Sigma-Aldrich
Tris(tert-butoxy)silanol, packaged for use in deposition systems
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%