  • Search Results
  • Extrinsic apoptosis is impeded by direct binding of the APL fusion protein NPM-RAR to TRADD.
Extrinsic apoptosis is impeded by direct binding of the APL fusion protein NPM-RAR to TRADD.
Molecular cancer research : MCR (2014-07-19)
Anuja Chattopadhyay, Brian L Hood, Thomas P Conrads, Robert L Redner
ABSTRACT
A subset of acute promyelocytic leukemia (APL) cases has been characterized by the t(5;17)(q35;q21) translocation variant, which fuses nucleophosmin (NPM) to retinoic acid receptor α (RARA). The resultant NPM-RAR fusion protein blocks myeloid differentiation and leads to a leukemic phenotype similar to that caused by the t(15;17)(q22;q21) PML-RAR fusion. The contribution of the N-terminal 117 amino acids of NPM contained within NPM-RAR has not been well studied. As a molecular chaperone, NPM interacts with a variety of proteins implicated in leukemogenesis. Therefore, a proteomic analysis was conducted to identify novel NPM-RAR-associated proteins. TNF receptor type I-associated DEATH domain protein (TRADD) was identified as a relevant binding partner for NPM-RAR. This interaction was validated by coprecipitation and colocalization analysis. Biologic assessment found that NPM-RAR expression impaired TNF-induced signaling through TRADD, blunting TNF-mediated activation of caspase-3 (CASP3) and caspase-8 (CASP8), to ultimately block apoptosis. This study identifies a novel mechanism through which NPM-RAR affects leukemogenesis.
Sigma-Aldrich
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in HEK 293 cells, suitable for cell culture
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
TNF-α human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)