  • Search Results
  • Stable expression and characterization of monomeric and dimeric recombinant hybrid-IgG/IgA immunoglobulins specific for Shiga toxin.
Stable expression and characterization of monomeric and dimeric recombinant hybrid-IgG/IgA immunoglobulins specific for Shiga toxin.
Biological & pharmaceutical bulletin (2014-07-06)
Koki Iwata, Kohta Kurohane, Katsuhiro Nakanishi, Masaki Miyake, Yasuyuki Imai
ABSTRACT
Antigen-specific immunoglobulin A (IgA) may be useful for preventing infectious diseases through passive immunization on the mucosal surface. We previously established mouse IgA and IgG monoclonal antibodies (mAbs) specific for the binding subunit of Shiga toxin 1 (Stx1B). We also developed a recombinant hybrid-IgG/IgA, in which variable regions from the IgG mAb were present. The binding activity of recombinant hybrid-IgG/IgA was verified by transient expression. Aiming at a constant supply, we established Chinese hamster ovary cells stably expressing monomeric or dimeric hybrid-IgG/IgA. The cDNAs encoding heavy and light chains were co-expressed for the monomeric hybrid-IgG/IgA, while those encoding heavy, light, and joining chains were co-expressed for the dimeric one. Serum-free culture supernatants of the cloned transfectants were subjected to size-exclusion chromatography. The elution patterns showed that the binding to immobilized Stx1B and the immunoblot signals of assembled immunoglobulins were correlated. In the transfectant for the dimeric hybrid-IgG/IgA, both monomers and dimers were observed. Size-exclusion chromatography enabled us to prepare a sample of the dimeric hybrid-IgG/IgA devoid of the monomeric one. The monomeric and dimeric forms of hybrid-IgG/IgA were prepared from the respective transfectants to examine the neutralization of Stx1. After pretreatment with monomeric or dimeric hybrid-IgG/IgA, the cytotoxicity of Stx1 toward Vero cells was abolished. Furthermore, the dimeric form was more than 10-fold more effective than the monomeric one in terms of toxin neutralization. These results suggest that the tetravalent feature of the binding sites of the dimeric hybrid-IgG/IgA contributes to the efficacy of toxin neutralization.
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, Animal Component-free
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, meets USP testing specifications, powder
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, powder, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)