  • Search Results
  • Bioassay-guided isolation of Poliovirus-inhibiting constituents from Zephyranthes candida.
Bioassay-guided isolation of Poliovirus-inhibiting constituents from Zephyranthes candida.
Pharmaceutical biology (2014-11-29)
Ogbole Omonike Oluyemisi, Ajaiyeoba Edith Oriabure, Adeniji J Adekunle, Kamdem Soup T Ramsay, Sajan Shyyaula, Muhammad Iqbal Choudhary
ABSTRACT
Plants of the Zephyranthes genus are globally used in folk medicine. In a previous study, Zephyranthes candida Linn. (Amaryllidaceae) was identified as having antiviral properties; this led to anti-poliovirus assay-guided isolation of compounds from crude methanol extract of the plant. Isolation of anti-poliovirus constituents from Z. candida. Active chloroform fraction from crude methanol extract of Z. candida (whole plant) was subjected to bioassay-guided fractionation; repeated column and preparative thin layer chromatography led to isolation of active compounds. Chemical structures were identified using spectroscopic techniques. Using serial two-fold dilution of maximum non-toxic concentration (MNTC) of each compound (0.0625-1 µg/mL for lycorine and 0.625-10 µg/mL for trisphaeridine and 7-hydroxy-3',4'-methylenedioxyflavan), the ability of extracts to inhibit viral-induced cell death in tissue culture was evaluated 72 h post-infection by the colorimetric method using MTT (3-[4,5-dimethylthiazol-2-yl]-2,5-diphenyltetrazolium bromide) dye. Regression analysis was used to determine 50% inhibitory concentration (IC50) and 50% cytotoxicity concentration (CC50), from which selective index (SI) was calculated. From the chloroform fraction, three compounds were isolated and identified, namely lycorine (1), trisphaeridine (2), and 7-hydroxy-3',4'-methylenedioxyflavan (3) as the anti-polioviral components. Lycorine was the most active, with an IC50 value of 0.058 µg/mL followed by trisphaeridine (2) with an IC50 of 0.1427 µg/mL, and 7-hydroxy-3',4'-methylenedioxyflavan (3) with an IC50 of 0.2384 µg/mL. The antipoliovirus activity of trisphaeridine (2) and 7-hydroxy-3',4'-methylenedioxyflavan (3) is established in this report; these compounds are of moderate toxicity and have very good SI. They could be a potential template for the development of a new antiviral agent.
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Pyridine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Pyridine, ≥99%
Supelco
Pyridine, Pharmaceutical Secondary Standard; Certified Reference Material