MilliporeSigma
  • Down-regulation of BTG3 promotes cell proliferation, migration and invasion and predicts survival in gastric cancer.
Down-regulation of BTG3 promotes cell proliferation, migration and invasion and predicts survival in gastric cancer.
Journal of cancer research and clinical oncology (2014-09-23)
X L Ren, X H Zhu, X M Li, Y L Li, J M Wang, P X Wu, Z B Lv, W H Ma, W T Liao, W Wang, Y Q Ding, L Liang
ABSTRACT
Gastric cancer (GC) is one of the most common malignancies in China. B-cell translocation gene 3 (BTG3) has been identified as a tumor suppressor in several tumors, but its role in GC remains unknown. This study aimed to detect the expression of BTG3 and its prognostic value in GC tissues and determine its function in the progression of GC. The expression of BTG3 was detected in GC cell lines and tissues by real-time RT-PCR, Western blot or immunohistochemistry. A series of in vitro and in vivo assays were performed to evaluate the effect of BTG3 on proliferation, migration and invasion of GC cells. B-cell translocation gene 3 was obviously down-regulated in GC tissues. Its expression was positively correlated with distant metastasis (P < 0.05). Patients with lower BTG3 expression had shorter overall survival time (P = 0.015). BTG3 suppressed the proliferation of GC cells in vitro and in vivo. It also inhibited migration and invasion of GC cells in vitro. Down-regulation of BTG3 is closely associated with proliferation, migration and invasion in GC. It may be a novel prognostic biomarker for GC patients.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard