  • Search Results
  • Lyn tyrosine kinase regulates androgen receptor expression and activity in castrate-resistant prostate cancer.
Lyn tyrosine kinase regulates androgen receptor expression and activity in castrate-resistant prostate cancer.
Oncogenesis (2014-08-19)
A Zardan, K M Nip, D Thaper, P Toren, S Vahid, E Beraldi, L Fazli, F Lamoureux, K M Gust, M E Cox, J L Bishop, A Zoubeidi
ABSTRACT
Castrate-resistant prostate cancer (CRPC) progression is a complex process by which prostate cells acquire the ability to survive and proliferate in the absence or under very low levels of androgens. Most CRPC tumors continue to express the androgen receptor (AR) as well as androgen-responsive genes owing to reactivation of AR. Protein tyrosine kinases have been implicated in supporting AR activation under castrate conditions. Here we report that Lyn tyrosine kinase expression is upregulated in CRPC human specimens compared with hormone naive or normal tissue. Lyn overexpression enhanced AR transcriptional activity both in vitro and in vivo and accelerated CRPC. Reciprocally, specific targeting of Lyn resulted in a decrease of AR transcriptional activity in vitro and in vivo and prolonged time to castration. Mechanistically, we found that targeting Lyn kinase induces AR dissociation from the molecular chaperone Hsp90, leading to its ubiquitination and proteasomal degradation. This work indicates a novel mechanism of regulation of AR stability and transcriptional activity by Lyn and justifies further investigation of the Lyn tyrosine kinase as a therapeutic target for the treatment of CRPC.Oncogenesis (2014) 3, e115; doi:10.1038/oncsis.2014.30; published online 18 August 2014.
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Sigma-Aldrich
DAPI, for nucleic acid staining