  • Search Results
  • Identification of novel adipokines in the joint. Differential expression in healthy and osteoarthritis tissues.
Identification of novel adipokines in the joint. Differential expression in healthy and osteoarthritis tissues.
PloS one (2015-04-09)
Javier Conde, Morena Scotece, Vanessa Abella, Rodolfo Gómez, Verónica López, Rosa Villar, Miguel Hermida, Jesús Pino, Juan Jesús Gómez-Reino, Oreste Gualillo
ABSTRACT
Emerging data suggest that several metabolic factors, released mainly by white adipose tissue (WAT) and joint tissues, and collectively named adipokines, might have a role in the pathophysiology of OA. Recently, novel adipokines such as SERPINE2, WISP2, GPNMB and ITIH5 have been identified in WAT. The main goal of this study was to analyse the expression of these novel adipokines in synovium, infrapatellar fat pad and chondrocytes and to compare the expression of these molecules in healthy and OA tissues. Synovial tissues, infrapatellar fat pad and chondrocytes were obtained from 36 OA patients (age 52-85; mean BMI 28.9) who underwent total knee replacement surgery. Healthy synovial tissues and infrapatellar fat pad were obtained from 15 traumatic knee patients (age 23-53; mean BMI 23.5). mRNA and protein expression were determined by qRT-PCR and western blot analysis respectively. All the novel adipokines, matter of our study, are expressed in OA synovium, infrapatellar fat pad and chondrocytes. Moreover, we detected a differential expression of SERPINE2 and ITIH5 in OA synovial tissues as compared to healthy samples. Finally, we also observed an increased expression of WISP2 in OA infrapatellar fat pad in comparison to healthy controls. In this study we demonstrated for the first time the expression of four novel adipokines in different joint tissues and how these molecules are differentially expressed in healthy and OA joint tissues.
Product Description
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Supelco
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl